2-(Methylthio)naphthalene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl(naphthalen-2-yl)sulphane

340119-5G 340119-25G
77492-808EA 145.23 USD
77492-808 77502-646
2-(Methylthio)naphthalene ≥95%
2-(Methylthio)naphthalene

Precaución: For research use only

Formula: C₁₁H₁₀S
MW: 174.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051615
CAS Number: 7433-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR