6-Amino-3-methyluracil ≥95%

Supplier: AOBChem USA

Warning

332678-5G 332678-25G
77492-666EA 72.1 USD
77492-666 77502-514
6-Amino-3-methyluracil ≥95%
6-Amino-3-methyluracil

Precaución: For research use only

Formula: C₅H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD02091062
CAS Number: 21236-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR