1-(4-Isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

331193-5G 331193-25G
77492-628EA 163.77 USD
77492-628 77502-482
1-(4-Isopropoxyphenyl)ethanone ≥95%
1-(4-Isopropoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD02254121
CAS Number: 4074-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR