3-Bromo-5-chloro-2-fluoroaniline ≥95%
Supplier: AOBChem USA
|
Warning
|
28791-5G
28791-25G
77491-246EA
65.92
USD
77491-246
77501-252
3-Bromo-5-chloro-2-fluoroaniline ≥95%
3-Bromo-5-chloro-2-fluoroaniline
Precaución: For research use only
Formula:
C₆H₄BrClFN MW: 224.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19382423 CAS Number: 1269232-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...