1,2-Dimethylnaphthalene ≥97%

Supplier: AOBChem USA

Warning

272308-5G 272308-25G
77490-964EA 146.26 USD
77490-964 77501-016
1,2-Dimethylnaphthalene ≥97%
1,2-Dimethylnaphthalene

Precaución: For research use only

Formula: C₁₂H₁₂
MW: 156.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004035
CAS Number: 573-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR