(4-Chlorophenyl)(4-nitrophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-4'-nitrodiphenyl sulphide, 4-(4-Chlorophenylthio)nitrobenzene

251392-5G 251392-25G
77490-514EA 67.98 USD
77490-514 77500-660
(4-Chlorophenyl)(4-nitrophenyl)sulfane ≥95%
(4-(4-Chlorophenyl)(4-nitrophenyl)sulfane)(4-nitrophenyl)sulphane

Precaución: For research use only

Formula: C₁₂H₈ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD00043613
CAS Number: 21969-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR