2-Acetylamino-3-bromo-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

24179-5G 24179-25G
77490-278EA 152.44 USD
77490-278 77500-454
2-Acetylamino-3-bromo-5-methylpyridine ≥97%
2-Acetylamino-3-bromo-5-methylpyridine

Precaución: For research use only

Formula: C₈H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06798091
CAS Number: 142404-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR