3-(4'-Fluorobenzyloxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-((4-Fluorobenzyl)oxy)phenyl)boronic acid

22904-5G 22904-25G
77490-080EA 109.18 USD
77490-080 77500-318
3-(4'-Fluorobenzyloxy)phenylboronic acid ≥95%
3-(4'-Fluorobenzyloxy)phenylboronic acid

Precaución: For research use only

Formula: C₁₃H₁₂BFO₃
MW: 246.0420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705238
CAS Number: 1072952-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR