1-Bromo-4-(2-chloroethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-β-chlorophenetol, 4-Bromo-ω-chlorophenetol, 4-Bromophenyl 2-chloroethyl ether

21934-5G 21934-25G
77489-990EA 181.28 USD
77489-990 77500-242
1-Bromo-4-(2-chloroethoxy)benzene ≥95%
1-Bromo-4-(2-chloroethoxy)benzene

Precaución: For research use only

Formula: C₈H₈BrClO
MW: 235.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000969
CAS Number: 55162-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR