3'-Ethylacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3'-Ethylacetophenone, m-Ethylacetophenone, 1-(3-Ethylphenyl)ethanone

209943-5G 209943-25G
77489-734EA 164.8 USD
77489-734 77500-038
3'-Ethylacetophenone ≥95%
3'-Ethylacetophenone

Precaución: For research use only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009660
CAS Number: 22699-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR