4,6-Dichloropyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

20884-5G 20884-25G
77489-720EA 100.94 USD
77489-720 77500-022
4,6-Dichloropyridine-3-carbaldehyde ≥97%
4,6-Dichloropyridine-3-carbaldehyde

Precaución: For research use only

Formula: C₆H₃Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD12025485
CAS Number: 1060811-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR