4,4-Dimethylcyclohexanol ≥96%

Supplier: AOBChem USA

Warning

203048-5G 203048-25G
77489-656EA 69.01 USD
77489-656 77499-976
4,4-Dimethylcyclohexanol ≥96%
4,4-Dimethylcyclohexanol

Precaución: For research use only

Formula: C₈H₁₆O
MW: 128.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00101954
CAS Number: 932-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR