2,6-Dichloro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

20177-5G 20177-25G
77489-594EA 60.77 USD
77489-594 77499-918
2,6-Dichloro-4-iodophenol ≥97%
2,6-Dichloro-4-iodophenol

Precaución: For research use only

Formula: C₆H₃Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD00466446
CAS Number: 34074-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR