3-(3-Bromophenyl)-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(3-Bromophenyl)-1H-pyrazole

198173-5G 198173-25G
77489-560EA 71.07 USD
77489-560 77499-886
3-(3-Bromophenyl)-1H-pyrazole ≥95%
3-(3-Bromophenyl)-1H-pyrazole

Precaución: For research use only

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01940433
CAS Number: 149739-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR