3-Bromo-4,5-dimethylnitrobenzene ≥95%

Supplier: AOBChem USA

Warning

19369-5G 19369-25G
77489-494EA 87.55 USD
77489-494 77499-840
3-Bromo-4,5-dimethylnitrobenzene ≥95%
3-Bromo-4,5-dimethylnitrobenzene

Precaución: For research use only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD12547850
CAS Number: 22162-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR