3-Bromo-4-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

192509-5G 192509-25G
77489-476EA 91.67 USD
77489-476 77499-826
3-Bromo-4-isopropylbenzoic acid ≥95%
3-Bromo-4-isopropylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11110860
CAS Number: 99070-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR