5-Cyano-3-pyridinylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18890-5G 18890-25G
77489-372EA 100.94 USD
77489-372 77499-730
5-Cyano-3-pyridinylboronic acid ≥97%
5-Cyano-3-pyridinylboronic acid

Precaución: For research use only

Formula: C6H5BN2O2
MW: 147.9300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374882
CAS Number: 497147-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR