5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole ≥97%

Supplier: AOBChem USA

Warning

182843-5G 182843-25G
77489-138EA 82.4 USD
77489-138 77499-570
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole ≥97%
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole

Precaución: For research use only

Formula: C₇H₄BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD04972651
CAS Number: 887267-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR