2-Phenyl-2-pentanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Phenylpentan-2-ol

182353-5G 182353-25G
77489-086EA 83.43 USD
77489-086 77499-540
2-Phenyl-2-pentanol ≥97%
2-Phenyl-2-pentanol

Precaución: For research use only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046592
CAS Number: 4383-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR