dibenzo[b,d]furan-2-carbaldehyde ≥96%

Supplier: AOBChem USA

Warning

Synonyms: dibenzo[b,d]furan-2-carbaldehyde, Dibenzofuran-2-carboxaldehyde

181609-5G 181609-25G
77489-048EA 146.26 USD
77489-048 77499-500
dibenzo[b,d]furan-2-carbaldehyde ≥96%
dibenzo[b,d]furan-2-carbaldehyde

Precaución: For research use only

Formula: C₁₃H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00136272
CAS Number: 5397-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR