4-Methoxy-2-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

18016-5G 18016-25G
77488-970EA 166.86 USD
77488-970 77499-460
4-Methoxy-2-(trifluoromethoxy)benzoic acid ≥97%
4-Methoxy-2-(trifluoromethoxy)benzoic acid

Precaución: For research use only

Formula: C₉H₇F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD06660285
CAS Number: 886502-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR