4-Ethylthioanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Ethyl-4-(methylthio)benzene

173177-5G 173177-25G
77488-814EA 182.31 USD
77488-814 77499-322
4-Ethylthioanisole ≥95%
4-Ethylthioanisole

Precaución: For research use only

Formula: C₉H₁₂S
MW: 152.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789171
CAS Number: 31218-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR