4'-Bromo-3'-fluoroacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-3-fluoro--acetophenone
17075-5G
17075-25G
77488-790EA
60.77
USD
77488-790
77499-298
4'-Bromo-3'-fluoroacetophenone ≥97%
4'-Bromo-3'-fluoroacetophenone
Precaución: For research use only
Formula:
C₈H₆BrFO MW: 217.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00466241 CAS Number: 304445-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...