Ethyl 7-bromo-1-benzothiophene-2-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Ethyl 7-bromo-1-benzothiophene-2-carboxylate, Ethyl 7-bromobenzo[b]thiophene-2-carboxylate
169568-5G
169568-25G
77488-714EA
99.91
USD
77488-714
77499-232
Ethyl 7-bromo-1-benzothiophene-2-carboxylate ≥97%
Ethyl 7-bromo-1-benzothiophene-2-carboxylate
Precaución: For research use only
Formula:
C₁₁H₉BrO₂S Storage Temperature: Ambient |
MDL Number:
MFCD17168278 CAS Number: 1355171-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...