2-Fluoro-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

16834-5G 16834-25G
77488-668EA 60.77 USD
77488-668 77499-204
2-Fluoro-6-(trifluoromethyl)benzamide ≥97%
2-Fluoro-6-(trifluoromethyl)benzamide

Precaución: For research use only

Formula: C₈H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD00083555
CAS Number: 144851-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR