1-(3-Fluorophenyl)propanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Fluorophenyl)propanol, alpha-Ethyl-3-fluorobenzyl alcohol

166930-5G 166930-25G
77488-520EA 109.18 USD
77488-520 77499-128
1-(3-Fluorophenyl)propanol ≥95%
1-(3-Fluorophenyl)propanol

Precaución: For research use only

Formula: C₉H₁₁FO
MW: 154.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062380
CAS Number: 701-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR