4-Bromo-2-methoxy-5-methylaniline ≥96%

Supplier: AOBChem USA

Warning

157801-5G 157801-25G
77488-084EA 60.77 USD
77488-084 77498-784
4-Bromo-2-methoxy-5-methylaniline ≥96%
4-Bromo-2-methoxy-5-methylaniline

Precaución: For research use only

Formula: C₈H₁₀BrNO
MW: 216.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547798
CAS Number: 873980-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR