3-Chloro-2-iodobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-2-iodo-3-(trifluoromethyl)benzene

15552-5G 15552-25G
77488-028EA 164.8 USD
77488-028 77498-730
3-Chloro-2-iodobenzotrifluoride ≥97%
3-Chloro-2-iodobenzotrifluoride

Precaución: For research use only

Formula: C₇H₃ClF₃I
MW: 306.4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00673987
CAS Number: 203626-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR