Ethyl 2-chloro-5-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

154709-5G 154709-25G
77488-016EA 164.8 USD
77488-016 77498-706
Ethyl 2-chloro-5-methylnicotinate ≥97%
Ethyl 2-chloro-5-methylnicotinate

Precaución: For research use only

Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610952
CAS Number: 894074-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR