3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

14990-5G 14990-25G
77487-912EA 76.22 USD
77487-912 77498-612
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester ≥97%
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester

Precaución: For research use only

Formula: C₁₄H₁₈BNO₆
Storage Temperature: Ambient
MDL Number: MFCD09258774
CAS Number: 957061-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR