2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Isopropoxy-5-(trifluoromethyl)phenyl)boronic acid, 2-Isopropoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid

14428-5G 14428-25G
77487-798EA 111.24 USD
77487-798 77498-518
2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid ≥95%
2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: For research use only

Formula: C₁₀H₁₂BF₃O₃
MW: 248.0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363786
CAS Number: 850593-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR