3-(Benzyloxy)-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

14252-5G 14252-25G
77487-780EA 127.72 USD
77487-780 77498-496
3-(Benzyloxy)-4-methylbenzaldehyde ≥95%
3-(Benzyloxy)-4-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD01632516
CAS Number: 1026868-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR