3-Chloro-4-(trifluoromethyl)phenol ≥95%

Supplier: AOBChem USA

Warning

131048-5G 131048-25G
77487-498EA 109.18 USD
77487-498 77498-288
3-Chloro-4-(trifluoromethyl)phenol ≥95%
3-Chloro-4-(trifluoromethyl)phenol

Precaución: For research use only

Formula: C₇H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD11849933
CAS Number: 37900-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR