(5-Methylthiophen-2-yl)methanamine ≥95%

Supplier: AOBChem USA

Warning

127049-5G 127049-25G
77487-356EA 170.98 USD
77487-356 77498-158
(5-Methylthiophen-2-yl)methanamine ≥95%
(5-Methylthiophen-2-yl)methanamine

Precaución: For research use only

Formula: C₆H₉NS
MW: 127.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00965306
CAS Number: 104163-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR