2-Bromo-4-chloro-6-(trifluoromethoxy)aniline ≥96%

Supplier: AOBChem USA

Warning

126940-5G 126940-25G
77487-342EA 160.68 USD
77487-342 77498-146
2-Bromo-4-chloro-6-(trifluoromethoxy)aniline ≥96%
2-Bromo-4-chloro-6-(trifluoromethoxy)aniline

Precaución: For research use only

Formula: C₇H₄BrClF₃NO
MW: 290.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14706173
CAS Number: 1244949-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR