4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-[(Dimethylamino)methyl]benzeneboronic acid pinacol ester, Dimethyl-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzyl]amine, N,N-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine

11884-5G 11884-25G
77487-112EA 126.69 USD
77487-112 77497-952
4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester ≥97%
4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester

Precaución: For research use only

Formula: C₁₅H₂₄BNO₂
MW: 261.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971190
CAS Number: 878197-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR