2-Chloro-4-(trifluoromethyl)nicotinic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethyl)-3-pyridinecarboxylic acid

11505-5G 11505-25G
77486-888EA 127.72 USD
77486-888 77497-752
2-Chloro-4-(trifluoromethyl)nicotinic acid ≥95%
2-Chloro-4-(trifluoromethyl)nicotinic acid

Precaución: For research use only

Formula: C₇H₃ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD08458125
CAS Number: 590371-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR