2-Bromo-5-fluoro-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-5-fluoropyridin-4-yl)boronic acid, 2-Bromo-5-fluoropyridine-4-boronic acid, 2-Bromo-5-fluoro-4-pyridinylboronic acid

11086-5G 11086-25G
77486-824EA 140.08 USD
77486-824 77497-678
2-Bromo-5-fluoro-4-pyridineboronic acid ≥97%
2-Bromo-5-fluoro-4-pyridineboronic acid

Precaución: For research use only

Formula: C₅H₄BBrFNO₂
MW: 219.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973353
CAS Number: 1072951-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR