3'-Chloro-4'-hydroxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Chloro-4-hydroxyphenyl)ethanone, 3-Chloro-4-hydroxyacetophenone, 1-(3-Chloro-4-hydroxyphenyl)ethan-1-one

105568-5G 105568-25G
77486-740EA 88.58 USD
77486-740 77497-570
3'-Chloro-4'-hydroxyacetophenone ≥97%
3'-Chloro-4'-hydroxyacetophenone

Precaución: For research use only

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00663621
CAS Number: 2892-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR