Protect It! Surge Suppressors, Tripp Lite
Supplier: Tripp Lite
STRIKER SUPER-7
89079-148EA 36.61 USD
89079-148 89079-150
Protect It! Surge Suppressors, Tripp Lite
Surge Suppressors
Surge suppressors feature seven outlets, EMI/RFI noise filtering, diagnostic LEDs, and plastic housing construction
STRIKER model provides economical surge protection for workstations, personal computers, telecom systems, and other sensitive electronics. Features 1.8m (6') power cord, one designated transformer plug outlet that doesn't block the remaining outlets, 15A circuit breaker, fail-safe thermal fusing that protects against fire and other damage in the event of extreme, extended overvoltage or surge conditions. Single row of outlets accepts up to four transformers. Built-in LED confirms suppressor "protection present" (green) status.
SUPER7 model provides surge protection for personal computers, CAD/CAM/CAE workstations, network nodes, fileservers, point-of-sale terminals, audio/visual devices, and more. Features 2.1m (7') power cord with a space-saving right-angle plug, and space for three large transformers without covering the remaining outlets. Includes a single row of five outlets plus two side-mounted outlets. Accepts up to five transformers. Built-in LED confirms suppressor "protection present" (green) and "grounded" (red) status.
Protection modes include full normal mode (H-N) and common mode (N-G/H-G) line surge suppression. Keyhole supports allow wall mounting.
Certificaciones: UL 1449 1998 Rev. (AC suppression), UL 1363 (power tap), and cUL listed. Exceed IEEE 587 category A
Información para pedidos: Manufacturer's lifetime warranty.