Provide premium surge, spike, and line noise protection of personal computers with peripherals, network and CAD/CAM/CAE workstations, telecommunications systems, point-of-sale equipment, audio/video and home theater systems in home, office, and industrial settings.
Robust surge suppression with sine-wave tracking protects against the heaviest of surge conditions, reducing 6000V test surges to a harmless level of less than 35V. Multi-component suppression circuits combine toroidal chokes, ferrite rod-core inductors, HF/VHF capacitors, and multiple layers of metal oxide varistors into exclusive isolated bank filters that remove EMI/RFI interference and even noise from other pieces of connected equipment.
Four, six, or eight high-quality duplex outlets are arranged in two, three, or four filter banks to prevent system crashes, reboots, and common performance problems when noisy peripherals are powered on or off. Three LED indicators confirm suppressor operation and site wiring status, as well as warn of suppressor or electrical fault conditions. Lighted power switch confirms power on/off status. Widely spaced outlets accommodate 2-4 large transformer plugs without blocking outlets. Outlet covers safely seal off unused outlets. Includes all-metal strip format with space-saving angled input plug. Keyhole mounting tabs allow permanent installation. Color: gray with black receptacles.
Six-outlet model includes two-phone line protection for modems, fax machines, and cordless phones (four-wire, two-line, RJ11 jacks).
Certificaciones: UL 1449, UL 1283, UL 1363, and cUL listed. Exceeds IEEE 587 category A
Información para pedidos: Manufacturer's lifetime warranty. NEMA 5-15R wall receptacle required for electrical service.