Nalgene® Packaging Bottles, High-Density Polyethylene, Narrow Mouth, without Screw Caps, Thermo Scientific
Supplier: Thermo Fisher Scientific
362089-0001 362089-0002 362089-0004 362089-0016 362089-0032
16060-106CS 528 USD
16060-106 16060-108 16060-110 16060-114 16060-116
Nalgene® Packaging Bottles, High-Density Polyethylene, Narrow Mouth, without Screw Caps, Thermo Scientific
Bottles Plastic Bottles
These economical bottles are made from high-density polyethylene.
  • Guaranteed leakproof
Single-use, break-resistant natural bottles allow easy viewing of liquid level.
Información para pedidos: For polypropylene caps, see 16060-026 series.