Disposable Tank Liners, Thermo Scientific
Supplier: LIFE TECHNOLOGIES CORP
SH30399.03 SH30647.03 SH30646.03 SH30647.01 SH30647.04 SH30647.05 SH30399.02 SH30646.02 SH30647.02 SH30646.01
82010-008EA 610.62 USD
82010-008 82010-050 82013-422 82013-424 82013-430 82013-432 82013-322 82013-420 82013-426 82013-418
Disposable Tank Liners, Thermo Scientific
Bags Media Bags
These disposable tank liners offer flexibility, reliability, and convenience throughout manufacturing processes.
  • Tank liners are designed for use with standard outer containers
Tank liners are manufactured in a variety of sizes and formulations designed to suit a wide array of laboratory applications. Designed to fit conical drums or cylindrical containers, liners are available in top- or bottom-dispensing designs, irradiated or nonirradiated, and constructed of either 4 or 9 mil film.
Bottom-dispensing tank liners are equipped with a bottom port for easy drainage and absolute fluid recovery. They are also equipped with a 12.7 mm quick-connect insert line.
Due to their single-use, disposable design, these tank liners eliminate the risk of cross-contamination and significantly reduce cleaning and turnaround time. Film used in the production of these tank liners is highly compatible with a wide range of media, process liquids, and buffers.
Información para pedidos: Tank liners are designed for use with standard outer containers.