Propyl acetate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Acetic acid propyl ester

A0044-500ML A0044-25ML
TCA0044-500MLEA 44.93 USD
TCA0044-500ML TCA0044-25ML
Propyl acetate ≥98.0%
Propyl acetate

CAS Number: 109-60-4
MDL Number: MFCD00009372
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 13
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Acetic Acid Propyl Ester

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 101 °C
Melting Pt: –95 °C
Density: 0.89 (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009372
CAS Number: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3,II
Merck Index: 12,08026
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0eaebeff28b43b4aedf5e51294f0d1124852258&vid=ac0e3b294c3785ed77e04b3284b870382130b6a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR