2-Fluoro-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%

Supplier: AOBChem USA

Warning

99949-1G 99949-500MG 99949-5G 99949-25G
77473-984EA 123.6 USD
77473-984 77486-594 77497-220 77506-102
2-Fluoro-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%
2-Fluoro-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: For research use only

Formula: C₁₄H₁₉BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD18383765
CAS Number: 1221188-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR