4-(4-pentylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99448-1G 99448-500MG 99448-5G 99448-25G
77473-976EA 415.09 USD
77473-976 77486-586 77497-212 77506-094
4-(4-pentylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(4-pentylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₆H₂₄O₂
MW: 248.361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34776039
CAS Number: 2918833-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR