4-(2-Isopropoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

99358-1G 99358-500MG 99358-5G 99358-25G
77473-964EA 393.46 USD
77473-964 77486-574 77497-200 77506-084
4-(2-Isopropoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2-Isopropoxyphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₄H₂₀O₃
MW: 236.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24834451
CAS Number: 1527700-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR