1-Bromo-2-(diethoxymethyl)-3,4-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

99337-1G 99337-500MG 99337-5G 99337-25G
77473-960EA 415.09 USD
77473-960 77486-570 77497-196 77506-080
1-Bromo-2-(diethoxymethyl)-3,4-difluorobenzene ≥95%
1-Bromo-2-(diethoxymethyl)-3,4-difluorobenzene

Precaución: For research use only

Formula: C₁₁H₁₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34775969
CAS Number: 2901099-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR