2-Bromo-6-(dimethoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

99314-1G 99314-500MG 99314-5G 99314-25G
77473-956EA 340.93 USD
77473-956 77486-566 77497-192 77506-076
2-Bromo-6-(dimethoxymethyl)pyridine ≥95%
2-Bromo-6-(dimethoxymethyl)pyridine

Precaución: For research use only

Formula: C₈H₁₀BrNO₂
MW: 232.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847413
CAS Number: 128507-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR