2-Bromo-1,4-difluoro-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

99265-1G 99265-500MG 99265-5G 99265-25G
77473-952EA 415.09 USD
77473-952 77486-562 77497-188 77506-072
2-Bromo-1,4-difluoro-3-(methoxymethoxy)benzene ≥97%
2-Bromo-1,4-difluoro-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34775943
CAS Number: 2015216-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR